Learn More
Abnova™ Human C4B Partial ORF (NP_000343, 774 a.a. - 873 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_000343 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 721 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16185414
|
Abnova™
H00000721-Q02.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16175414
|
Abnova™
H00000721-Q02.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes the basic form of complement factor 4, part of the classical activation pathway. The protein is expressed as a single chain precursor which is proteolytically cleaved into a trimer of alpha, beta, and gamma chains prior to secretion. The trimer provides a surface for interaction between the antigen-antibody complex and other complement components. The alpha chain may be cleaved to release C4 anaphylatoxin, a mediator of local inflammation. Deficiency of this protein is associated with systemic lupus erythematosus. This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6. Varying haplotypes of this gene cluster exist, such that individuals may have 1, 2, or 3 copies of this gene. In addition, this gene exists as a long form and a short form due to the presence or absence of a 6.4 kb endogenous HERV-K retrovirus in intron 9. [provided by RefSeq]
Sequence: VRSFFPENWLWRVETVDRFQILTLWLPDSLTTWEIHGLSLSKTKGLCVATPVQLRVFREFHLHLRLPMSVRRFEQLELRPVLYNYLDKNLTVSVHVSPVESpecifications
NP_000343 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C4A/C4A13/C4A91/C4B1/C4B12/C4B2/C4B3/C4B5/C4F/CH/CO4/CPAMD3/FLJ60561/MGC164979 | |
C4B | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
721 | |
C4B (Human) Recombinant Protein (Q02) | |
VRSFFPENWLWRVETVDRFQILTLWLPDSLTTWEIHGLSLSKTKGLCVATPVQLRVFREFHLHLRLPMSVRRFEQLELRPVLYNYLDKNLTVSVHVSPVE | |
RUO | |
C4B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |