missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human Ca1 Partial Orf (Aah27890.1, 12 A.A.-150 A.A.) Recombinant Protein With Gst Tag At N-Terminal.
Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH27890.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 759 |
Name | CA1 (Human) Recombinant Protein (Q01) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16013705
|
Abnova™
H00000759-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16023705
|
Abnova™
H00000759-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Good Folding
- Excellent amino acids labeling
Immunogen Sequence: NGPEQWSKLYPIANGNNQSPVDIKTSETKHDTSLKPISVSYNPATAKEIINVGHSFHVNFEDNDNRSVLKGGPFSDSYRLFQFHFHWGSTNEHGSEHTVDGVKYSAELHVAHWNSAKYSSLAEAASKADGLAVIGVLMK
Specifications
AAH27890.1 | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer. | |
CA1 (Human) Recombinant Protein (Q01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Car1 | |
CA1 | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
759 | |
Glutathione Sepharose 4 Fast Flow | |
NGPEQWSKLYPIANGNNQSPVDIKTSETKHDTSLKPISVSYNPATAKEIINVGHSFHVNFEDNDNRSVLKGGPFSDSYRLFQFHFHWGSTNEHGSEHTVDGVKYSAELHVAHWNSAKYSSLAEAASKADGLAVIGVLMK | |
RUO | |
CA1 | |
GST |
For Research Use Only