missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CABYR Full-length ORF (NP_619585.1, 1 a.a. - 379 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00026256-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16118232

  • 4515.00 SEK / 10µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

To reach fertilization competence, spermatozoa undergo a series of morphological and molecular maturational processes, termed capacitation, involving protein tyrosine phosphorylation and increased intracellular calcium. The protein encoded by this gene localizes to the principal piece of the sperm flagellum in association with the fibrous sheath and exhibits calcium-binding when phosphorylated during capacitation. A pseudogene on chromosome 3 has been identified for this gene. Transcript variants of this gene encode multiple protein isoforms. An additional transcript and isoform has not been fully characterized. [provided by RefSeq]

Sequence: MISSKPRLVVPYGLKTLLEGISRAVLKTNPSNINQFAAAYFQELTMYRGNTTMDIKDLVKQFHQIKVEKWSEGTTPQKKLECLKEPGKTSVESKVPTQMEKSTDTDEDNVTRTEYSDKTTQFPSVYAVPGTEQTEAVGGLSSKPATPKTTTPPSSPPPTAVSPEFAYVPADPAQLAAQMLAMATSERGQPPPCSNMWTLYCLTDKNQQGHPSPPPAPGPFPQATLYLPNPKDPQFQQHPPKVTFPTYVMGDTKKTSAPPFILVGSNVQEAQGWKPLPGHAVVSQSDVLRYVAMQVPIAVPADEKYQKHTLSPQNANPPSGQDVPRPKSPVFLSVAFPVEDVAKKSSGSGDKCAPFGSYGIAGEVTVTTAHKRRKAETEN
Specifications

Specifications

NP_619585.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
67.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CBP86/FSP-2/FSP2/MGC9117
CABYR
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
26256
CABYR (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MISSKPRLVVPYGLKTLLEGISRAVLKTNPSNINQFAAAYFQELTMYRGNTTMDIKDLVKQFHQIKVEKWSEGTTPQKKLECLKEPGKTSVESKVPTQMEKSTDTDEDNVTRTEYSDKTTQFPSVYAVPGTEQTEAVGGLSSKPATPKTTTPPSSPPPTAVSPEFAYVPADPAQLAAQMLAMATSERGQPPPCSNMWTLYCLTDKNQQGHPSPPPAPGPFPQATLYLPNPKDPQFQQHPPKVTFPTYVMGDTKKTSAPPFILVGSNVQEAQGWKPLPGHAVVSQSDVLRYVAMQVPIAVPADEKYQKHTLSPQNANPPSGQDVPRPKSPVFLSVAFPVEDVAKKSSGSGDKCAPFGSYGIAGEVTVTTAHKRRKAETEN
RUO
CABYR
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers