Learn More
Abnova™ Human CACNG3 Partial ORF (NP_006530, 199 a.a. - 297 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_006530 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10368 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16182416
|
Abnova™
H00010368-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16172416
|
Abnova™
H00010368-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
L-type calcium channels are composed of five subunits. The protein encoded by this gene represents one of these subunits, gamma, and is one of several gamma subunit proteins. It is an integral membrane protein that is thought to stabilize the calcium channel in an inactive (closed) state. This protein is similar to the mouse stargazin protein, mutations in which have been associated with absence seizures, also known as petit-mal or spike-wave seizures. This gene is a member of the neuronal calcium channel gamma subunit gene subfamily of the PMP-22/EMP/MP20 family. This gene is a candidate gene for a familial infantile convulsive disorder with paroxysomal choreoathetosis. [provided by RefSeq]
Sequence: IYIEKHQQLRAKSHSEFLKKSTFARLPPYRYRFRRRSSSRSTEPRSRDLSPISKGFHTIPSTDISMFTLSRDPSKITMGTLLNSDRDHAFLQFHNSTPKSpecifications
NP_006530 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Cacng2 | |
CACNG3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10368 | |
CACNG3 (Human) Recombinant Protein (Q01) | |
IYIEKHQQLRAKSHSEFLKKSTFARLPPYRYRFRRRSSSRSTEPRSRDLSPISKGFHTIPSTDISMFTLSRDPSKITMGTLLNSDRDHAFLQFHNSTPK | |
RUO | |
CACNG3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |