missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CALB2 Full-length ORF (NP_009018.1, 1 a.a. - 179 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4055.00 SEK - 6150.00 SEK
Specifications
Name | CALB2 (Human) Recombinant Protein (P01) |
---|---|
Recombinant | Yes |
Purification Method | Glutathione Sepharose 4 Fast Flow |
For Use With (Application) | Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) |
Molecular Weight | 47.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16115947
|
Abnova™
H00000794-P01.25UG |
25 ug |
6150.00 SEK
25µg |
Estimated Shipment: 28-07-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
16105947
|
Abnova™
H00000794-P01.10UG |
10 ug |
4055.00 SEK
10µg |
Estimated Shipment: 28-07-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Description
This gene encodes an intracellular calcium-binding protein belonging to the troponin C superfamily. Members of this protein family have six EF-hand domains which bind calcium. Three alternatively spliced transcript variants that encode different proteins have been described. [provided by RefSeq]
Sequence: MAGPQQQPPYLHLAELTASQFLEIWKHFDADGNGYIEGKELENFFQELEKARKGSGMMSKSDNFGEKMKEFMQKYDKNSDGKIEMAELAQILPTEENFLLCFRQHVGSSAEFMEAWRKYDTDRSGYIEANELKGFLSDLLKKANRPYDEPKLQEYTQTILRMFDLNGDGKLGLSEMSRASpecifications
CALB2 (Human) Recombinant Protein (P01) | |
Glutathione Sepharose 4 Fast Flow | |
47.1kDa |
Yes | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |