Learn More
Abnova™ Human CAPN7 Partial ORF (NP_055111.1, 714 a.a. - 813 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023473-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Calpains are ubiquitous, well-conserved family of calcium-dependent, cysteine proteases. The calpain proteins are heterodimers consisting of an invariant small subunit and variable large subunits. The large subunit possesses a cysteine protease domain, and both subunits possess calcium-binding domains. Calpains have been implicated in neurodegenerative processes, as their activation can be triggered by calcium influx and oxidative stress. The function of the protein encoded by this gene is not known. An orthologue has been found in mouse but it seems to diverge from other family members. The mouse orthologue is thought to be calcium independent with protease activity. [provided by RefSeq]
Sequence: NPIYQFHIEKTGPLLIELRGPRQYSVGFEVVTVSTLGDPGPHGFLRKSSGDYRCGFCYLELENIPSGIFNIIPSTFLPKQEGPFFLDFNSIIPIKITQLQSpecifications
NP_055111.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NPIYQFHIEKTGPLLIELRGPRQYSVGFEVVTVSTLGDPGPHGFLRKSSGDYRCGFCYLELENIPSGIFNIIPSTFLPKQEGPFFLDFNSIIPIKITQLQ | |
RUO | |
CAPN7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23473 | |
CAPN7 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CALPAIN7/FLJ36423/PALBH | |
CAPN7 | |
Recombinant | |
wheat germ expression system |