Learn More
Abnova™ Human CASC4 Full-length ORF (NP_612432.2, 1 a.a. - 436 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_612432.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 113201 |
Molecular Weight (g/mol) | 75.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16176983
|
Abnova™
H00113201-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16166983
|
Abnova™
H00113201-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The increased expression level of this gene is associated with HER-2/neu proto-oncogene overexpression. Amplification and resulting overexpression of this proto-oncogene are found in approximately 30% of human breast and 20% of human ovarian cancers. Further characterization of the product of this gene may yield insight into the fundamental biology and pathogenetic effects of HER-2/neu overexpression in human breast and ovarian cancer cells. Alternatively spliced variants encoding different isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MVGFGANRRAGRLPSLVLVVLLVVIVVLAFNYWSISSRHVLLQEEVAELQGQVQRTEVARGRLEKRNSDLLLLVDTHKKQIDQKEADYGRLSSRLQAREGLGKRCEDDKVKLQNNISYQMADIHHLKEQLAELRQEFLRQEDQLQDYRKNNTYLVKRLEYESFQCGQQMKELRAQHEENIKKLADQFLEEQKQETQKIQSNDGKELDINNQVVPKNIPKVAENVADKNEEPSSNHIPHGKEQIKRGGDAGMPGIEENDLAKVDDLPPALRKPPISVSQHESHQAISHLPTGQPLSPNMPPDSHINHNGNPGTSKQNPSSPLQRLIPGSNLDSEPRIQTDILKQATKDRVSDFHKLKQSRFFDENESPVDPQHGSKLADYNGDDGNVGEYEADKQAELAYNEEEDGDGGEEDVQDDEERELQMDPADYGKQHFNDVLSpecifications
NP_612432.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
75.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MVGFGANRRAGRLPSLVLVVLLVVIVVLAFNYWSISSRHVLLQEEVAELQGQVQRTEVARGRLEKRNSDLLLLVDTHKKQIDQKEADYGRLSSRLQAREGLGKRCEDDKVKLQNNISYQMADIHHLKEQLAELRQEFLRQEDQLQDYRKNNTYLVKRLEYESFQCGQQMKELRAQHEENIKKLADQFLEEQKQETQKIQSNDGKELDINNQVVPKNIPKVAENVADKNEEPSSNHIPHGKEQIKRGGDAGMPGIEENDLAKVDDLPPALRKPPISVSQHESHQAISHLPTGQPLSPNMPPDSHINHNGNPGTSKQNPSSPLQRLIPGSNLDSEPRIQTDILKQATKDRVSDFHKLKQSRFFDENESPVDPQHGSKLADYNGDDGNVGEYEADKQAELAYNEEEDGDGGEEDVQDDEERELQMDPADYGKQHFNDVL | |
RUO | |
CASC4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
113201 | |
CASC4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp459F1927/H63/MGC74708 | |
CASC4 | |
Recombinant | |
wheat germ expression system |