missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CASP5 Partial ORF (NP_004338, 309 a.a. - 418 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000838-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135654

  • 6845.00 SEK / 25µg
Estimated Shipment: 02-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. Overexpression of the active form of this enzyme induces apoptosis in fibroblasts. Max, a central component of the Myc/Max/Mad transcription regulation network important for cell growth, differentiation, and apoptosis, is cleaved by this protein; this process requires Fas-mediated dephosphorylation of Max. The expression of this gene is regulated by interferon-gamma and lipopolysaccharide. Several alternatively spliced transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq]

Sequence: VRDSPASLAVISSQSSENLEADSVCKIHEEKDFIAFCSSTPHNVSWRDRTRGSIFITELITCFQKYSCCCHLMEIFRKVQKSFEVPQAKAQMPTIERATLTRDFYLFPGN
Specifications

Specifications

NP_004338
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VRDSPASLAVISSQSSENLEADSVCKIHEEKDFIAFCSSTPHNVSWRDRTRGSIFITELITCFQKYSCCCHLMEIFRKVQKSFEVPQAKAQMPTIERATLTRDFYLFPGN
RUO
CASP5
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
838
CASP5 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ICE(rel)III/ICEREL-III/ICH-3/MGC141966
CASP5
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers