Learn More
Novus Biologicals™ Human Caspase-4 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand:  Novus Biologicals™ NBP2-75025
Product Code. 18323160
  • 7771.94 SEK / 1 set
Explore more special offers
Description
Description
The Human Caspase-4 ELISA Kit (Colorimetric) measures human Caspase-4 in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 - 10 ng/mL
This kit recognizes Human CASP4 in samples. No significant cross-reactivity or interference between Human CASP4 and analogues was observed.
Biological Fluids, Plasma, Serum
837
Human
Storage of components varies. See protocol for specific instructions.
0.10 ng/mL
ELISA Kit (Colorimetric)
ELISA
CASP4
1 Kit
Caspase-4