Learn More
Invitrogen™ eBioscience™ Annexin V, APC, Recombinant Protein
Recombinant Protein
Brand:  Invitrogen™ BMS306APC-20
Additional Details : Weight : 0.23750kg
 View more versions of this product
Product Code. 15501917
  • 896.00 SEK / 20 tests
Estimated Shipment date
from Supplier 22-01-2018
Add to basket
Description
Description
Annexins are a family of calcium-dependent phospholipid-binding proteins, which bindto phosphatidylserine (PS) to identify apoptotic cells. In healthy cells, PS is predominantlylocated along the cytosolic side of the plasma membrane. Upon initiation of apoptosis,PS loses its asymmetric distribution in the phospholipid bilayer and translocates tothe extracellular membrane, which is detectable with fluorescently labeled AnnexinV. In early stages of apoptosis, the plasma membrane excludes viability dyes such aspropidium iodide (PI) and 7-AAD, therefore cells which display only Annexin V staining(PI/7-AAD negative) are in early stages of apoptosis. During late-stage apoptosis, loss ofcell membrane integrity allows Annexin V binding to cytosolic PS, as well as cell uptakeof PI and 7-AAD. Annexin V staining, paired with 7-AAD or PI is widely used to identifyapoptotic stages by flow cytometry.
Specifications
Specifications
Flow Cytometric Analysis
35.8kDa
SDS-gel electrophoresis and reverse phase HPLC
20 tests
Annexin V-APC is stable at 2-8deg;C as indicated on the label. Do not freeze!
E. Coli
Annexin V exhibits anti-phospholipase activity and binds to phosphatidylserine (PS).
APC
Solution containing 50mM TRIS, 100mM NaCl, 1% BSA, 0.02% Sodium Azide, pH 7.4
Annexin V
>89%
E. coli
RUO
Lipocortin V, Endonexin II, Calphobindin I
Recombinant Protein