missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Gibco™ Human CCL25 (TECK), Animal-Free Recombinant Protein, PeproTech®
Beskrivning
Recombinant Human TECK (CCL25) is a 14.2 kDa protein containing 127 amino acid residues, including the four conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Chemokines are likely to play an important role in regulating the trafficking of developing T cells within the thymus. Chemokine C-C thymus expressed chemokine (TECK), also designated chemokine ligand 25 (CCL25), small induc-ible cytokine A25, chemokine b-15 or CK b-15, is expressed predominantly in thymic dendritic cells, thymic epithelial cells and in the small intestine. TECK, a CCR9 ligand, has suppressive activity against immature subsets of myeloid progenitors which have been stimulated to proliferate by multiple growth factors. TECK delivers signals through CCR9, which is important for the nav-igation of developing thymocytes. Bone marrow pre-pro-B cells and cells capable of generating pro-B colonies in the presence of interleukin-7 and Flt-3 ligand migrate to TECK, a response lost in later stages of B cell development.
Specifikationer
Specifikationer
| Tillträdesnummer | O15444 |
| För användning med (applikation) | Functional Assay |
| Formulering | protein with no preservative |
| Gen-ID (Entrez) | 6370 |
| Molekylvikt (g/mol) | 14.2 kDa |
| Namn | Human CCL25 (TECK), Animal-Free |
| Kvantitet | 100 μg |
| Källa | E. coli |
| Regulatorisk status | RUO |
| Endotoxinkoncentration | <0.1 EU/ μg |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?