Learn More
Abnova™ Human CCNG1 Partial ORF (AAH00196, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000900-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The eukaryotic cell cycle is governed by cyclin-dependent protein kinases (CDKs) whose activities are regulated by cyclins and CDK inhibitors. The protein encoded by this gene is a member of the cyclin family and contains the cyclin box. The encoded protein lacks the protein destabilizing (PEST) sequence that is present in other family members. Transcriptional activation of this gene can be induced by tumor protein p53. Two transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq]
Sequence: MIEVLTTTDSQKLLHQLNALLEQESRCQPKVCGLRLIESAHDNGLRMTARLRDFEVKDLLSLTQFFGFDTETFSLAVNLLDRFLSKMKVQPKHLGCVGLSCFYLAVKSIESpecifications
AAH00196 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MIEVLTTTDSQKLLHQLNALLEQESRCQPKVCGLRLIESAHDNGLRMTARLRDFEVKDLLSLTQFFGFDTETFSLAVNLLDRFLSKMKVQPKHLGCVGLSCFYLAVKSIE | |
RUO | |
CCNG1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
900 | |
CCNG1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CCNG | |
CCNG1 | |
Recombinant | |
wheat germ expression system |