Learn More
Abnova™ Human CCRN4L Partial ORF (NP_036250, 64 a.a. - 152 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00025819-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is highly similar to Nocturnin, a gene identified as a circadian clock regulated gene in Xenopus laevis. This protein and Nocturnin protein share similarity with the C-terminal domain of a yeast transcription factor, carbon catabolite repression 4 (CCR4). The mRNA abundance of a similar gene in mouse has been shown to exhibit circadian rhythmicity, which suggests a role for this protein in clock function or as a circadian clock effector. [provided by RefSeq]
Sequence: VCSMGTGTSRLYSALAKTLNSSAASQHPEYLVSPDPEHLEPIDPKELLEECRAVLHTRPPRFQRDFVDLRTDCPSTHPPIRVMQWNILASpecifications
NP_036250 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VCSMGTGTSRLYSALAKTLNSSAASQHPEYLVSPDPEHLEPIDPKELLEECRAVLHTRPPRFQRDFVDLRTDCPSTHPPIRVMQWNILA | |
RUO | |
CCRN4L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
25819 | |
CCRN4L (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CCR4L/MGC142054/MGC142060/MGC4120817/MGC78549/NOC | |
CCRN4L | |
Recombinant | |
wheat germ expression system |