missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CD226 Partial ORF (NP_006557, 21 a.a. - 127 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_006557 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10666 |
Molecular Weight (g/mol) | 37.51kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16123006
|
Abnova™
H00010666-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16113006
|
Abnova™
H00010666-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a glycoprotein expressed on the surface of NK cells, platelets, monocytes and a subset of T cells. It is a member of the Ig-superfamily containing 2 Ig-like domains of the V-set. The protein mediates cellular adhesion of platelets and megakaryocytic cells to vascular endothelial cells. The protein also plays a role in megakaryocytic cell maturation. [provided by RefSeq]
Sequence: VLWHTSVPFAENMSLECVYPSMGILTQVEWFKIGTQQDSIAIFSPTHGMVIRKPYAERVYFLNSTMASNNMTLFFRNASEDDVGYYSCSLYTYPQGTWQKVIQVVQSSpecifications
NP_006557 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.51kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DNAM-1/DNAM1/PTA1/TLiSA1 | |
CD226 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10666 | |
CD226 (Human) Recombinant Protein (Q01) | |
VLWHTSVPFAENMSLECVYPSMGILTQVEWFKIGTQQDSIAIFSPTHGMVIRKPYAERVYFLNSTMASNNMTLFFRNASEDDVGYYSCSLYTYPQGTWQKVIQVVQS | |
RUO | |
CD226 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |