Läs mer
R&D Systems™ Human CD79A (NP_001774) VersaClone cDNA
Beskrivning
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
Beställningsinfo
Ships at ambient temperature
Specifikationer
Specifikationer
| Gen | CD79A |
| Tillträdesnummer | NP_001774 |
| Koncentration | 10 μg at 0.2 μg/μL |
| Formulering | cDNA is provided in 10mM Tris-Cl, pH 8.329 |
| Infoga typ | 694bp |
| Gen-ID (Entrez) | 973 |
| Gene Alias | CD79A, CD79a antigen, CD79A antigen (immunoglobulin-associated alpha), CD79a molecule, immunoglobulin-associated alpha, IGAB-cell antigen receptor complex-associated protein alpha chain, Ig-alpha, MB1, MB-1, MB-1 membrane glycoprotein, Membrane-bound immunoglobulin-associated protein, Surface IgM-associated protein |
| Innehåll och lagring | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Art | Human |
| Kvantitet | 10 μg |
For Reasearch Use Only. Not for use in Humans.
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.