Learn More
Abnova™ Human CDH3 Full-length ORF (AAH41846.1, 1 a.a. - 829 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH41846.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1001 |
Molecular Weight (g/mol) | 117.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16182971
|
Abnova™
H00001001-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16172971
|
Abnova™
H00001001-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is a classical cadherin from the cadherin superfamily. The encoded protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. This gene is located in a six-cadherin cluster in a region on the long arm of chromosome 16 that is involved in loss of heterozygosity events in breast and prostate cancer. In addition, aberrant expression of this protein is observed in cervical adenocarcinomas. Mutations in this gene have been associated with congential hypotrichosis with juvenile macular dystrophy. [provided by RefSeq]
Sequence: MGLPRGPLASLLLLQVCWLQCAASEPCRAVFREAEVTLEAGGAEQEPGQALGKVFMGCPGQEPALFSTDNDDFTVRNGETVQERRSLKERNPLKIFPSKRILRRHKRDWVVAPISVPENGKGPFPQRLNQLKSNKDRDTKIFYSITGPGADSPPEGVFAVEKETGWLLLNKPLDREEIAKYELFGHAVSENGASVEDPMNISIIVTDQNDHKPKFTQDTFRGSVLEGVLPGTSVMQMTATDEDDAIYTYNGVVAYSIHSQEPKDPHDLMFTIHRSTGTISVISSGLDREKVPEYTLTIQATDMDGDGSTTTAVAVVEILDANDNAPMFDPQKYEAHVPENAVGHEVQRLTVTDLDAPNSPAWRATYLIMGGDDGDHFTITTHPESNQGILTTRKGLDFEAKNQHTLYVEVTNEAPFVLKLPTSTATIVVHVEDVNEAPVFVPPSKVVEVQEGIPTGEPVCVYTAEDPDKENQKISYRILRDPAGWLAMDPDSGQVTAVGTLDREDEQFVRNNIYEVMVLAMDNGSPPTTGTGTLLLTLIDVNDHGPVPEPRQITICNQSPVRQVLNITDKDLSPHTSPFQAQLTDDSDIYWTAEVNEEGDTVVLSLKKFLKQDTYDVHLSLSDHGNKEQLTVIRATVCDCHGHVETCPGPWKGGFILPVLGAVLALLFLLLVLLLLVRKKRKIKEPLLLPEDDTRDNVFYYGEEGGGEEDQDYDITQLHRGLEARPEVVLRNDVAPTIIPTPMYRPRPANPDEIGNFIIENLKAANTDPTAPPYDTLLVFDYEGSGSDAASLSSLTSSASDQDQDYDYLNEWGSRFKKLADMYGGGEDDSpecifications
AAH41846.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
117.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGLPRGPLASLLLLQVCWLQCAASEPCRAVFREAEVTLEAGGAEQEPGQALGKVFMGCPGQEPALFSTDNDDFTVRNGETVQERRSLKERNPLKIFPSKRILRRHKRDWVVAPISVPENGKGPFPQRLNQLKSNKDRDTKIFYSITGPGADSPPEGVFAVEKETGWLLLNKPLDREEIAKYELFGHAVSENGASVEDPMNISIIVTDQNDHKPKFTQDTFRGSVLEGVLPGTSVMQMTATDEDDAIYTYNGVVAYSIHSQEPKDPHDLMFTIHRSTGTISVISSGLDREKVPEYTLTIQATDMDGDGSTTTAVAVVEILDANDNAPMFDPQKYEAHVPENAVGHEVQRLTVTDLDAPNSPAWRATYLIMGGDDGDHFTITTHPESNQGILTTRKGLDFEAKNQHTLYVEVTNEAPFVLKLPTSTATIVVHVEDVNEAPVFVPPSKVVEVQEGIPTGEPVCVYTAEDPDKENQKISYRILRDPAGWLAMDPDSGQVTAVGTLDREDEQFVRNNIYEVMVLAMDNGSPPTTGTGTLLLTLIDVNDHGPVPEPRQITICNQSPVRQVLNITDKDLSPHTSPFQAQLTDDSDIYWTAEVNEEGDTVVLSLKKFLKQDTYDVHLSLSDHGNKEQLTVIRATVCDCHGHVETCPGPWKGGFILPVLGAVLALLFLLLVLLLLVRKKRKIKEPLLLPEDDTRDNVFYYGEEGGGEEDQDYDITQLHRGLEARPEVVLRNDVAPTIIPTPMYRPRPANPDEIGNFIIENLKAANTDPTAPPYDTLLVFDYEGSGSDAASLSSLTSSASDQDQDYDYLNEWGSRFKKLADMYGGGEDD | |
RUO | |
CDH3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
1001 | |
CDH3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDHP/HJMD/PCAD | |
CDH3 | |
Recombinant | |
wheat germ expression system |