Learn More
Novus Biologicals™ Human CEBP gamma ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand:  Novus Biologicals™ NBP2-75219
Product Code. 18322281
  • 7771.94 SEK / 1 set
Explore more special offers
Description
Description
The Human CEBP gamma ELISA Kit (Colorimetric) measures human CEBP gamma in serum, plasma and other biological fluids.
Specifications
Specifications
3.13 - 200 ng/mL
This kit recognizes Human C/EBP(gamma) in samples. No significant cross-reactivity or interference between Human C/EBP(gamma) and analogues was observed.
Biological Fluids, Plasma, Serum
1054
Human
Cell Cycle and Replication
CEBP gamma
1.88 ng/mL
ELISA Kit (Colorimetric)
ELISA
CEBPG
1 Kit
Storage of components varies. See protocol for specific instructions.