Learn More
Novus Biologicals™ Human CEBP gamma ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand: Novus Biologicals™ NBP2-75219
This item is not returnable.
View return policy
Description
The Human CEBP gamma ELISA Kit (Colorimetric) measures human CEBP gamma in serum, plasma and other biological fluids.Specifications
3.13 - 200 ng/mL | |
This kit recognizes Human C/EBP(gamma) in samples. No significant cross-reactivity or interference between Human C/EBP(gamma) and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
1054 | |
Human | |
Cell Cycle and Replication | |
CEBP gamma |
1.88 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
CEBPG | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. |