missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CENPE Partial ORF (NP_001804, 309 a.a. - 419 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifica
Accession Number | NP_001804 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1062 |
Molecular Weight (g/mol) | 37.95kDa |
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
16173111
|
Abnova™
H00001062-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16183111
|
Abnova™
H00001062-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descrizione
Centrosome-associated protein E is a kinesin-like motor protein that accumulates in the G2 phase of the cell cycle. Unlike other centrosome-associated proteins, it is not present during interphase and first appears at the centromere region of chromosomes during prometaphase. CENPE is proposed to be one of the motors responsible for mammalian chromosome movement and/or spindle elongation. [provided by RefSeq]
Sequence: TPVSFDETLTALQFASTAKYMKNTPYVNEVSTDEALLKRYRKEIMDLKKQLEEVSLETRAQAMEKDQLAQLLEEKDLLQKVQNEKIENLTRMLVTSSSLTLQQELKAKRKRSpecifica
NP_001804 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.95kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIF10 | |
CENPE | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
1062 | |
CENPE (Human) Recombinant Protein (Q01) | |
TPVSFDETLTALQFASTAKYMKNTPYVNEVSTDEALLKRYRKEIMDLKKQLEEVSLETRAQAMEKDQLAQLLEEKDLLQKVQNEKIENLTRMLVTSSSLTLQQELKAKRKR | |
RUO | |
CENPE | |
Wheat Germ (in vitro) | |
GST | |
Liquid |