missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CEP2 Partial ORF (AAH01433, 141 a.a. - 240 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH01433 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 11190 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16146342
|
Abnova™
H00011190-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16136342
|
Abnova™
H00011190-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a core centrosomal protein required for centriole-centriole cohesion during interphase of the cell cycle. The encoded protein dissociates from the centrosomes when parental centrioles separate at the beginning of mitosis. The protein associates with and is phosphorylated by NIMA-related kinase 2, which is also associated with the centrosome. [provided by RefSeq]
Sequence: LTVDWSRARDELMRKESQWQMEQEFFKGYLKGEHGRLLSLWREVVTFRRHFLEMKSATDRDLMELKAEHVRLSGSLLTCCLRLTVGAQSREPNGSGRMDGSpecifications
AAH01433 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C-NAP1/CEP2/CNAP1/MGC88542 | |
CEP250 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
11190 | |
CEP2 (Human) Recombinant Protein (Q01) | |
LTVDWSRARDELMRKESQWQMEQEFFKGYLKGEHGRLLSLWREVVTFRRHFLEMKSATDRDLMELKAEHVRLSGSLLTCCLRLTVGAQSREPNGSGRMDG | |
RUO | |
CEP250 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |