missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CEP57 Full-length ORF (NP_055494.2, 1 a.a. - 500 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009702-P02.25ug
Additional Details : Weight : 0.00010kg
Description
Translokin binds basic fibroblast growth factor (FGF2; MIM 134920) and mediates its nuclear translocation and mitogenic activity (Bossard et al., 2003 [PubMed 12717444]).[supplied by OMIM]
Sequence: MAAASVSAASGSHLSNSFAEPSRSNGSMVRHSSSPYVVYPSDKPFLNSDLRRSPSKPTLAYPESNSRAIFSALKNLQDKIRRLELERIQAEESVKTLSRETIEYKKVLDEQIQERENSKNEESKHNQELTSQLLAAENKCNLLEKQLEYMRNMIKHAEMERTSVLEKQVSLERERQHDQTHVQSQLEKLDLLEQEYNKLTTMQALAEKKMQELEAKLHEEEQERKRMQAKAAELQTGLETNRLIFEDKATPCVPNARRIKKKKSKPPEKKSSRNYFGAQPHYRLCLGDMPFVAGKSTSPSHAVVANVQLVLHLMKQHSKALCNDRVINSIPLAKQVSSRGGKSKKLSVTPPSSNGINEELSEVLQTLQDEFGQMSFDHQQLAKLIQESPTVELKDKLECELEALVGRMEAKANQITKVRKYQAQLEKQKLEKQKKELKATKKTLDEERNSSSRSGITGTTNKKDFMKLRPGEKRRKNLQLLKDMQSIQNSLQSSSLCWDYSpecifications
NP_055494.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
80.63kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0092/PIG8/TSP57 | |
CEP57 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9702 | |
CEP57 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAASVSAASGSHLSNSFAEPSRSNGSMVRHSSSPYVVYPSDKPFLNSDLRRSPSKPTLAYPESNSRAIFSALKNLQDKIRRLELERIQAEESVKTLSRETIEYKKVLDEQIQERENSKNEESKHNQELTSQLLAAENKCNLLEKQLEYMRNMIKHAEMERTSVLEKQVSLERERQHDQTHVQSQLEKLDLLEQEYNKLTTMQALAEKKMQELEAKLHEEEQERKRMQAKAAELQTGLETNRLIFEDKATPCVPNARRIKKKKSKPPEKKSSRNYFGAQPHYRLCLGDMPFVAGKSTSPSHAVVANVQLVLHLMKQHSKALCNDRVINSIPLAKQVSSRGGKSKKLSVTPPSSNGINEELSEVLQTLQDEFGQMSFDHQQLAKLIQESPTVELKDKLECELEALVGRMEAKANQITKVRKYQAQLEKQKLEKQKKELKATKKTLDEERNSSSRSGITGTTNKKDFMKLRPGEKRRKNLQLLKDMQSIQNSLQSSSLCWDY | |
RUO | |
CEP57 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |