missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CFC1 Partial ORF (NP_115934.1, 27 a.a. - 93 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055997-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138576

  • 6845.00 SEK / 25µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the epidermal growth factor (EGF)- Cripto, Frl-1, and Cryptic (CFC) family. EGF-CFC family member proteins share a variant EGF-like motif, a conserved cysteine-rich domain, and a C-terminal hydrophobic region. These proteins play key roles in intercellular signaling pathways during vertebrate embryogenesis. Mutations in this gene can cause autosomal visceral heterotaxy. This protein is involved in left-right asymmetric morphogenesis during organ development. [provided by RefSeq]

Sequence: QREKHNGGREEVTKVATQKHRQSPLNWTSSHFGEVTGSAEGWGPEEPLPYSRAFGEGASARPRCCRN
Specifications

Specifications

NP_115934.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.11kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QREKHNGGREEVTKVATQKHRQSPLNWTSSHFGEVTGSAEGWGPEEPLPYSRAFGEGASARPRCCRN
RUO
CFC1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
55997
CFC1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CRYPTIC/FLJ77897/HTX2/MGC133213
CFC1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers