Learn More
Abnova™ Human CHAF1B Full-length ORF (NP_005432.1, 1 a.a. - 559 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_005432.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8208 |
Molecular Weight (g/mol) | 87.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16124688
|
Abnova™
H00008208-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16114688
|
Abnova™
H00008208-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chromatin assembly factor I (CAF-I) is required for the assembly of histone octamers onto newly-replicated DNA. CAF-I is composed of three protein subunits, p50, p60, and p150. The protein encoded by this gene corresponds to the p60 subunit and is required for chromatin assembly after replication. The encoded protein is differentially phosphorylated in a cell cycle-dependent manner. In addition, it is normally found in the nucleus except during mitosis, when it is released into the cytoplasm. This protein is a member of the WD-repeat HIR1 family and may also be involved in DNA repair. [provided by RefSeq]
Sequence: MKVITCEIAWHNKEPVYSLDFQHGTAGRIHRLASAGVDTNVRIWKVEKGPDGKAIVEFLSNLARHTKAVNVVRFSPTGEILASGGDDAVILLWKVNDNKEPEQIAFQDEDEAQLNKENWTVVKTLRGHLEDVYDICWATDGNLMASASVDNTAIIWDVSKGQKISIFNEHKSYVQGVTWDPLGQYVATLSCDRVLRVYSIQKKRVAFNVSKMLSGIGAEGEARSYRMFHDDSMKSFFRRLSFTPDGSLLLTPAGCVESGENVMNTTYVFSRKNLKRPIAHLPCPGKATLAVRCCPVYFELRPVVETGVELMSLPYRLVFAVASEDSVLLYDTQQSFPFGYVSNIHYHTLSDISWSSDGAFLAISSTDGYCSFVTFEKDELGIPLKEKPVLNMRTPDTAKKTKSQTHRGSSPGPRPVEGTPASRTQDPSSPGTTPPQARQAPAPTVIRDPPSITPAVKSPLPGPSEEKTLQPSSQNTKAHPSRRVTLNTLQAWSKTTPRRINLTPLKTDTPPSSVPTSVISTPSTEEIQSETPGDAQGSPPELKRPRLDENKGGTESLDPSpecifications
NP_005432.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
87.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MKVITCEIAWHNKEPVYSLDFQHGTAGRIHRLASAGVDTNVRIWKVEKGPDGKAIVEFLSNLARHTKAVNVVRFSPTGEILASGGDDAVILLWKVNDNKEPEQIAFQDEDEAQLNKENWTVVKTLRGHLEDVYDICWATDGNLMASASVDNTAIIWDVSKGQKISIFNEHKSYVQGVTWDPLGQYVATLSCDRVLRVYSIQKKRVAFNVSKMLSGIGAEGEARSYRMFHDDSMKSFFRRLSFTPDGSLLLTPAGCVESGENVMNTTYVFSRKNLKRPIAHLPCPGKATLAVRCCPVYFELRPVVETGVELMSLPYRLVFAVASEDSVLLYDTQQSFPFGYVSNIHYHTLSDISWSSDGAFLAISSTDGYCSFVTFEKDELGIPLKEKPVLNMRTPDTAKKTKSQTHRGSSPGPRPVEGTPASRTQDPSSPGTTPPQARQAPAPTVIRDPPSITPAVKSPLPGPSEEKTLQPSSQNTKAHPSRRVTLNTLQAWSKTTPRRINLTPLKTDTPPSSVPTSVISTPSTEEIQSETPGDAQGSPPELKRPRLDENKGGTESLDP | |
RUO | |
CHAF1B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
8208 | |
CHAF1B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAF-1/CAF-IP60/CAF1/CAF1A/CAF1P60/MPHOSPH7/MPP7 | |
CHAF1B | |
Recombinant | |
wheat germ expression system |