missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CHD3 Partial ORF (NP_001005273, 1654 a.a. - 1741 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001107-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16133161

  • 4515.00 SEK / 10µg
Estimated Shipment: 20-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the CHD family of proteins which are characterized by the presence of chromo (chromatin organization modifier) domains and SNF2-related helicase/ATPase domains. This protein is one of the components of a histone deacetylase complex referred to as the Mi-2/NuRD complex which participates in the remodeling of chromatin by deacetylating histones. Chromatin remodeling is essential for many processes including transcription. Autoantibodies against this protein are found in a subset of patients with dermatomyositis. Three alternatively spliced transcripts encoding different isoforms have been described. [provided by RefSeq]

Sequence: KPLDGQEHRERPEGETGDLGKREDVKGDRELRPGPRDEPRSNGRREEKTEKPRFMFNIADGGFTELHTLWQNEERAAISSGKLNEIWH
Specifications

Specifications

NP_001005273
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.42kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KPLDGQEHRERPEGETGDLGKREDVKGDRELRPGPRDEPRSNGRREEKTEKPRFMFNIADGGFTELHTLWQNEERAAISSGKLNEIWH
RUO
CHD3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
1107
CHD3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Mi-2a/Mi2-ALPHA/ZFH
CHD3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers