Learn More
Abnova™ Human CITED1 Partial ORF (NP_004134, 94 a.a. - 193 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004435-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the CREB-binding protein/p300-interacting transactivator with Asp/Glu-rich C-terminal domain (CITED) family of proteins. The encoded protein, also known as melanocyte-specific gene 1, may function as a transcriptional coactivator and may play a role in pigmentation of melanocytes. Alternatively spliced transcript variants have been described. [provided by RefSeq]
Sequence: SMQLQKLNSQYQGMAAATPGQPGEAGPLQNWDFGAQAGGAESLSPSAGAQSPAIIDSDPVDEEVLMSLVVELGLDRANELPELWLGQNEFDFTADFPSSCSpecifications
NP_004134 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SMQLQKLNSQYQGMAAATPGQPGEAGPLQNWDFGAQAGGAESLSPSAGAQSPAIIDSDPVDEEVLMSLVVELGLDRANELPELWLGQNEFDFTADFPSSC | |
RUO | |
CITED1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4435 | |
CITED1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MSG1 | |
CITED1 | |
Recombinant | |
wheat germ expression system |