missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CLCN6 Partial ORF (NP_001277.1, 770 a.a. - 868 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001185-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136404

  • 6845.00 SEK / 25µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The CLCN family of voltage-dependent chloride channel genes comprises nine members (CLCN1-7, Ka and Kb) which demonstrate quite diverse functional characteristics while sharing significant sequence homology. Chloride channel 6 and 7 belong to a subbranch of this family. Chloride channel 6 has four different alternatively spliced transcript variants. This gene is in close vicinity to two other kidney-specific chloride channel genes, CLCNKA and CLCNKB. [provided by RefSeq]

Sequence: PRLSYAEMAEDYPRYPDIHDLDLTLLNPRMIVDVTPYMNPSPFTVSPNTHVSQVFNLFRTMGLRHLPVVNAVGEIVGIITRHNLTYEFLQARLRQHYQT
Specifications

Specifications

NP_001277.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PRLSYAEMAEDYPRYPDIHDLDLTLLNPRMIVDVTPYMNPSPFTVSPNTHVSQVFNLFRTMGLRHLPVVNAVGEIVGIITRHNLTYEFLQARLRQHYQT
RUO
CLCN6
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
1185
CLCN6 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CLC-6/KIAA0046
CLCN6
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers