missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CLDN7 Full-length ORF (NP_001298.2, 1 a.a. - 211 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001366-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16146637

  • 6845.00 SEK / 25µg
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Claudins, such as CLDN7, are involved in the formation of tight junctions between epithelial cells. Tight junctions restrict lateral diffusion of lipids and membrane proteins, and thereby physically define the border between the apical and basolateral compartments of epithelial cells (Zheng et al., 2003 [PubMed 14502431]).[supplied by OMIM]

Sequence: MANSGLQLLGFSMALLGWVGLVACTAIPQWQMSSYAGDNIITAQAMYKGLWMDCVTQSTGMMSCKMYDSVLALSAALQATRALMVVSLVLGFLAMFVATMGMKCTRCGGDDKVKKARIAMGGGIIFIVAGLATLVACSWYGHQIVTDFYNPLIPTNIKYEFGPAIFIGWAGSALVILGGALLSCSCPGNESKAGYRAPRSYPKSNSSKEYV
Specifications

Specifications

NP_001298.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CEPTRL2/CPETRL2/Hs.84359/claudin-1
CLDN7
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
1366
CLDN7 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MANSGLQLLGFSMALLGWVGLVACTAIPQWQMSSYAGDNIITAQAMYKGLWMDCVTQSTGMMSCKMYDSVLALSAALQATRALMVVSLVLGFLAMFVATMGMKCTRCGGDDKVKKARIAMGGGIIFIVAGLATLVACSWYGHQIVTDFYNPLIPTNIKYEFGPAIFIGWAGSALVILGGALLSCSCPGNESKAGYRAPRSYPKSNSSKEYV
RUO
CLDN7
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers