Learn More
Abnova™ Human CLEC10A Partial ORF (NP_006335.2, 70 a.a. - 169 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010462-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may function as a cell surface antigen. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: VTLRTDFSNFTSNTVAEIQALTSQGSSLEETIASLKAEVEGFKQERQAVHSEMLLRVQQLVQDLKKLTCQVATLNNNGEEASTEGTCCPVNWVEHQDSCYSpecifications
NP_006335.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VTLRTDFSNFTSNTVAEIQALTSQGSSLEETIASLKAEVEGFKQERQAVHSEMLLRVQQLVQDLKKLTCQVATLNNNGEEASTEGTCCPVNWVEHQDSCY | |
RUO | |
CLEC10A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10462 | |
CLEC10A (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD301/CLECSF13/CLECSF14/HML/HML2 | |
CLEC10A | |
Recombinant | |
wheat germ expression system |