missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CLEC9A Full-length ORF (NP_997228.1, 1 a.a. - 241 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00283420-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16179853

  • 5030.41 SEK / 25µg

Estimated Shipment: 22-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CLEC9A is a group V C-type lectin-like receptor (CTLR) that functions as an activation receptor and is expressed on myeloid lineage cells (Huysamen et al., 2008 [PubMed 18408006]).[supplied by OMIM]

Sequence: MHEEEIYTSLQWDSPAPDTYQKCLSSNKCSGACCLVMVISCVFCMGLLTASIFLGVKLLQVSTIAMQQQEKLIQQERALLNFTEWKRSCALQMKYCQAFMQNSLSSAHNSSPCPNNWIQNRESCYYVSEIWSIWHTSQENCLKEGSTLLQIESKEEMDFITGSLRKIKGSYDYWVGLSQDGHSGRWLWQDGSSPSPGLLPAERSQSANQVCGYVKSNSLLSSNCSTWKYFICEKYALRSSV
Specifications

Specifications

NP_997228.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
53.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DNGR1/UNQ9341
CLEC9A
Yes
wheat germ expression system
Liquid
283420
CLEC9A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MHEEEIYTSLQWDSPAPDTYQKCLSSNKCSGACCLVMVISCVFCMGLLTASIFLGVKLLQVSTIAMQQQEKLIQQERALLNFTEWKRSCALQMKYCQAFMQNSLSSAHNSSPCPNNWIQNRESCYYVSEIWSIWHTSQENCLKEGSTLLQIESKEEMDFITGSLRKIKGSYDYWVGLSQDGHSGRWLWQDGSSPSPGLLPAERSQSANQVCGYVKSNSLLSSNCSTWKYFICEKYALRSSV
RUO
CLEC9A
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates