Learn More
Abnova™ Human CLIC5 Partial ORF (NP_058625.2, 91 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00053405-Q01.25ug
Additional Details : Weight : 0.00010kg
Descrizione
Chloride intracellular channels are involved in chloride ion transport within various subcellular compartments. CLIC5 specifically associates with the cytoskeleton of placenta microvilli.[supplied by OMIM]
Sequence: FLEETLTPEKYPKLAAKHRESNTAGIDIFSKFSAYIKNTKQQNNAALERGLTKALKKLDDYLNTPLPEEIDANTCGEDKGSRRKFLDGDELTLADCNLLPSpecifica
NP_058625.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FLEETLTPEKYPKLAAKHRESNTAGIDIFSKFSAYIKNTKQQNNAALERGLTKALKKLDDYLNTPLPEEIDANTCGEDKGSRRKFLDGDELTLADCNLLP | |
RUO | |
CLIC5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
53405 | |
CLIC5 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLIC5B/FLJ90663/MST130/MSTP130/dJ447E21.4 | |
CLIC5 | |
Recombinant | |
wheat germ expression system |