missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CLIP4 Full-length ORF (NP_078968.3, 1 a.a. - 705 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079745-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16184423

  • 5030.41 SEK / 25µg

Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MTIEDLPDFPLEGNPLFGRYPFIFSASDTPVIFSISAAPMPSDCEFSFFDPNDASCQEILFDPKTSVSELFAILRQWVPQVQQNIDIIGNEILKRGCNVNDRDGLTDMTLLHYTCKSGAHGIGDVETAVKFATQLIDLGADISLRSRWTNMNALHYAAYFDVPELIRVILKTSKPKDVDATCSDFNFGTALHIAAYNLCAGAVKCLLEQGANPAFRNDKGQIPADVVPDPVDMPLEMADAAATAKEIKQMLLDAVPLSCNISKAMLPNYDHVTGKAMLTSLGLKLGDRVVIAGQKVGTLRFCGTTEFASGQWAGIELDEPEGKNNGSVGKVQYFKCAPKYGIFAPLSKISKAKGRRKNITHTPSTKAAVPLIRSQKIDVAHVTSKVNTGLMTSKKDSASESTLSLPPGEELKTVTEKDVALLGSVSSCSSTSSLEHRQSYPKKQNAISSNKKTMSKSPSLSSRASAGLNSSATSTANNSRCEGELRLGERVLVVGQRLGTIRFFGTTNFAPGYWYGIELEKPHGKNDGSVGGVQYFSCSPRYGIFAPPSRVQRVTDSLDTLSEISSNKQNHSYPGFRRSFSTTSASSQKEINRRNAFSKSKAALRRSWSSTPTAGGIEGSVKLHEGSQVLLTSSNEMGTVRYVGPTDFASGIWLGLELRSAKGKNDGSVGDKRYFTCKPNHGVLVRPSRVTYRGINGSKLVDENC
Specifications

Specifications

NP_078968.3
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
102.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ21069/FLJ32705/RSNL2
CLIP4
Yes
wheat germ expression system
Liquid
79745
CLIP4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTIEDLPDFPLEGNPLFGRYPFIFSASDTPVIFSISAAPMPSDCEFSFFDPNDASCQEILFDPKTSVSELFAILRQWVPQVQQNIDIIGNEILKRGCNVNDRDGLTDMTLLHYTCKSGAHGIGDVETAVKFATQLIDLGADISLRSRWTNMNALHYAAYFDVPELIRVILKTSKPKDVDATCSDFNFGTALHIAAYNLCAGAVKCLLEQGANPAFRNDKGQIPADVVPDPVDMPLEMADAAATAKEIKQMLLDAVPLSCNISKAMLPNYDHVTGKAMLTSLGLKLGDRVVIAGQKVGTLRFCGTTEFASGQWAGIELDEPEGKNNGSVGKVQYFKCAPKYGIFAPLSKISKAKGRRKNITHTPSTKAAVPLIRSQKIDVAHVTSKVNTGLMTSKKDSASESTLSLPPGEELKTVTEKDVALLGSVSSCSSTSSLEHRQSYPKKQNAISSNKKTMSKSPSLSSRASAGLNSSATSTANNSRCEGELRLGERVLVVGQRLGTIRFFGTTNFAPGYWYGIELEKPHGKNDGSVGGVQYFSCSPRYGIFAPPSRVQRVTDSLDTLSEISSNKQNHSYPGFRRSFSTTSASSQKEINRRNAFSKSKAALRRSWSSTPTAGGIEGSVKLHEGSQVLLTSSNEMGTVRYVGPTDFASGIWLGLELRSAKGKNDGSVGDKRYFTCKPNHGVLVRPSRVTYRGINGSKLVDENC
RUO
CLIP4
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates