missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CMPK2 Partial ORF (NP_997198, 151 a.a. - 250 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_997198 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 129607 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16121767
|
Abnova™
H00129607-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16111767
|
Abnova™
H00129607-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Mitochondrial UMP-CMP kinase (EC 2.7.2.14) is a component of the salvage pathway for nucleotide synthesis. Other enzymes of the salvage pathway include thymidine kinase-2 (TK2; MIM 188250), deoxynucleotidase-2 (NT5M; MIM 605292), deoxyguanosine kinase (DGUOK; MIM 601465), adenylate kinase-2 (AK2; MIM 103020), adenylate kinase-3 (AK3; MIM 609290), adenylate kinase-3-like-1 (AK3L1; MIM 103030), and nucleoside diphosphate kinase (NME4; MIM 601818) (Xu et al., 2008 [PubMed 17999954]).[supplied by OMIM]
Sequence: GACQEAPRPHLGEFEADPRGQLWQRLWEVQDGRRLQVGCAQVVPVPEPPLHPVVPDLPSSVVFPDREAARAVLEECTSFIPEARAVLDLVDQCPKQIQKGSpecifications
NP_997198 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TYKi/UMP-CMPK2 | |
CMPK2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
129607 | |
CMPK2 (Human) Recombinant Protein (Q01) | |
GACQEAPRPHLGEFEADPRGQLWQRLWEVQDGRRLQVGCAQVVPVPEPPLHPVVPDLPSSVVFPDREAARAVLEECTSFIPEARAVLDLVDQCPKQIQKG | |
RUO | |
CMPK2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |