missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CMTM8 Full-length ORF (NP_849199.2, 1 a.a. - 173 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_849199.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 152189 |
Molecular Weight (g/mol) | 46kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16128723
|
Abnova™
H00152189-P02.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16138723
|
Abnova™
H00152189-P02.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. This gene is widely expressed in many tissues, but the exact function of the encoded protein is unknown. [provided by RefSeq]
Sequence: MEEPQRARSHTVTTTASSFAENFSTSSSSFAYDREFLRTLPGFLIVAEIVLGLLVWTLIAGTEYFRVPAFGWVMFVAVFYWVLTVFFLIIYITMTYTRIPQVPWTTVGLCFNGSAFVLYLSAAVVDASSVSPERDSHNFNSWAASSFFAFLVTICYAGNTYFSFIAWRSRTIQSpecifications
NP_849199.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
46kDa | |
Glutathione Sepharose 4 Fast Flow | |
MEEPQRARSHTVTTTASSFAENFSTSSSSFAYDREFLRTLPGFLIVAEIVLGLLVWTLIAGTEYFRVPAFGWVMFVAVFYWVLTVFFLIIYITMTYTRIPQVPWTTVGLCFNGSAFVLYLSAAVVDASSVSPERDSHNFNSWAASSFFAFLVTICYAGNTYFSFIAWRSRTIQ | |
RUO | |
CMTM8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
152189 | |
CMTM8 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CKLFSF8/CKLFSF8-V2 | |
CMTM8 | |
Recombinant | |
wheat germ expression system |