missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CMTM8 Full-length ORF (NP_849199.2, 1 a.a. - 173 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00152189-P02.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138723

  • 6845.00 SEK / 25µg
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. This gene is widely expressed in many tissues, but the exact function of the encoded protein is unknown. [provided by RefSeq]

Sequence: MEEPQRARSHTVTTTASSFAENFSTSSSSFAYDREFLRTLPGFLIVAEIVLGLLVWTLIAGTEYFRVPAFGWVMFVAVFYWVLTVFFLIIYITMTYTRIPQVPWTTVGLCFNGSAFVLYLSAAVVDASSVSPERDSHNFNSWAASSFFAFLVTICYAGNTYFSFIAWRSRTIQ
Specifications

Specifications

NP_849199.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
46kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CKLFSF8/CKLFSF8-V2
CMTM8
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
152189
CMTM8 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEEPQRARSHTVTTTASSFAENFSTSSSSFAYDREFLRTLPGFLIVAEIVLGLLVWTLIAGTEYFRVPAFGWVMFVAVFYWVLTVFFLIIYITMTYTRIPQVPWTTVGLCFNGSAFVLYLSAAVVDASSVSPERDSHNFNSWAASSFFAFLVTICYAGNTYFSFIAWRSRTIQ
RUO
CMTM8
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers