Learn More
Abnova™ Human CNKSR1 Partial ORF (NP_006305, 111 a.a. - 210 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010256-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is a necessary element in receptor tyrosine kinase pathways, possibly as a tyrosine phosphorylation target. It is involved in regulation of RAF in the MAPK pathway and may also play a role in a MAPK-independent pathway. [provided by RefSeq]
Sequence: VLCAAVELLHEADALLFWLSRYLFSHLNDFSACQEIRDLLEELSQVLHEDGPAAEKEGTVLRICSHVAGICHNILVCCPKELLEQKAVLEQVQLDSPLGLSpecifications
NP_006305 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VLCAAVELLHEADALLFWLSRYLFSHLNDFSACQEIRDLLEELSQVLHEDGPAAEKEGTVLRICSHVAGICHNILVCCPKELLEQKAVLEQVQLDSPLGL | |
RUO | |
CNKSR1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10256 | |
CNKSR1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CNK/CNK1/KSR | |
CNKSR1 | |
Recombinant | |
wheat germ expression system |