missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CNO Partial ORF (NP_060836, 108 a.a. - 217 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_060836 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55330 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16178076
|
Abnova™
H00055330-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16168076
|
Abnova™
H00055330-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This intronless gene encodes a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. A similar protein in mouse is a component of a protein complex termed biogenesis of lysosome-related organelles complex 1 (BLOC-1), and is a model for Hermansky-Pudlak syndrome. The encoded protein may play a role in intracellular vesicular trafficking. [provided by RefSeq]
Sequence: GDSSHVVSEGVPRIHAKAAEMRRIYSRIDRLEAFVRMVGGRVARMEEQVTKAEAELGTFPRAFKKLLHTMNVPSLFSKSAPSRPQQAGYEAPVLFRTEDYFPCCSERPQLSpecifications
NP_060836 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BCAS4L/FLJ11230 | |
CNO | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
55330 | |
CNO (Human) Recombinant Protein (Q01) | |
GDSSHVVSEGVPRIHAKAAEMRRIYSRIDRLEAFVRMVGGRVARMEEQVTKAEAELGTFPRAFKKLLHTMNVPSLFSKSAPSRPQQAGYEAPVLFRTEDYFPCCSERPQL | |
RUO | |
CNO | |
Wheat Germ (in vitro) | |
GST | |
Liquid |