Learn More
Abnova™ Human CNTN2 Partial ORF (NP_005067, 825 a.a. - 923 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006900-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. It may also be involved in glial tumorigenesis and may provide a potential target for therapeutic intervention. [provided by RefSeq]
Sequence: SSSEMNVTWEPVQQDMNGILLGYEIRYWKAGDKEAAADRVRTAGLDTSARVSGLHPNTKYHVTVRAYNRAGTGPASPSANATTMKPPPRRPPGNISWTFSpecifications
NP_005067 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SSSEMNVTWEPVQQDMNGILLGYEIRYWKAGDKEAAADRVRTAGLDTSARVSGLHPNTKYHVTVRAYNRAGTGPASPSANATTMKPPPRRPPGNISWTF | |
RUO | |
CNTN2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6900 | |
CNTN2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AXT/DKFZp781D102/FLJ42746/MGC157722/TAG-1/TAX/TAX1 | |
CNTN2 | |
Recombinant | |
wheat germ expression system |