missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human COG4 Partial ORF (NP_056201, 686 a.a. - 785 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_056201 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 25839 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16115156
|
Abnova™
H00025839-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16105156
|
Abnova™
H00025839-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Multiprotein complexes are key determinants of Golgi apparatus structure and its capacity for intracellular transport and glycoprotein modification. Several complexes have been identified, including the Golgi transport complex (GTC), the LDLC complex, which is involved in glycosylation reactions, and the SEC34 complex, which is involved in vesicular transport. These 3 complexes are identical and have been termed the conserved oligomeric Golgi (COG) complex, which includes COG4 (Ungar et al., 2002 [PubMed 11980916]).[supplied by OMIM]
Sequence: ELEKVVLKSTFNRLGGLQFDKELRSLIAYLTTVTTWTIRDKFARLSQMATILNLERVTEILDYWGPNSGPLTWRLTPAEVRQVLALRIDFRSEDIKRLRLSpecifications
NP_056201 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COD1/DKFZp586E1519 | |
COG4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
25839 | |
COG4 (Human) Recombinant Protein (Q01) | |
ELEKVVLKSTFNRLGGLQFDKELRSLIAYLTTVTTWTIRDKFARLSQMATILNLERVTEILDYWGPNSGPLTWRLTPAEVRQVLALRIDFRSEDIKRLRL | |
RUO | |
COG4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |