Learn More
Novus Biologicals™ Human COL1A2 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand:  Novus Biologicals™ NBP2-75844
Product Code. 18322921
  • 8267.60 SEK / 1 set
Explore more special offers
Description
Description
The Human COL1A2 ELISA Kit (Colorimetric) measures human COL1A2 in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 - 10 ng/mL
This kit recognizes Human COL1a2 in samples. No significant cross-reactivity or interference between Human COL1a2 and analogues was observed.
Biological Fluids, Plasma, Serum
1278
Human
Storage of components varies. See protocol for specific instructions.
0.10 ng/mL
ELISA Kit (Colorimetric)
ELISA
COL1A2
1 Kit
COL1A2