missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human COL9A3 Partial ORF (NP_001844, 280 a.a. - 338 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_001844 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1299 |
Molecular Weight (g/mol) | 32.23kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16146524
|
Abnova™
H00001299-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16136524
|
Abnova™
H00001299-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes one of the three alpha chains of type IX collagen, the major collagen component of hyaline cartilage. Type IX collagen, a heterotrimeric molecule, is usually found in tissues containing type II collagen, a fibrillar collagen. Mutations in this gene are associated with multiple epiphyseal dysplasia. [provided by RefSeq]
Sequence: GDLGRPGPKGTPGVAGPSGEPGMPGKDGQNGVPGLDGQKGEAGRNGAPGEKGPNGLPGLSpecifications
NP_001844 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.23kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DJ885L7.4.1/EDM3/FLJ90759/IDD/MED | |
COL9A3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
1299 | |
COL9A3 (Human) Recombinant Protein (Q01) | |
GDLGRPGPKGTPGVAGPSGEPGMPGKDGQNGVPGLDGQKGEAGRNGAPGEKGPNGLPGL | |
RUO | |
COL9A3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |