Learn More
Abnova™ Human CRIM1 Partial ORF (NP_057525, 36 a.a. - 145 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051232-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Motor neurons are among the earliest neurons to appear after the commencement of cell patterning and the beginning of cell differentiation. Differentiation occurs in a ventral-to-dorsal gradient and is mediated, at least in part, by the concentration of ventrally expressed Sonic hedgehog protein (SHH; MIM 600725). Dorsally expressed factors, such as members of the bone morphogenic protein (e.g., BMP4; MIM 112262) and transforming growth factor-beta (e.g., TGFB1; MIM 190180) families, can repress the induction of these neurons. CRIM1 may interact with growth factors implicated in motor neuron differentiation and survival (Kolle et al., 2000 [PubMed 10642437]).[supplied by OMIM]
Sequence: VCLPCDESKCEEPRNCPGSIVQGVCGCCYTCASQRNESCGGTFGIYGTCDRGLRCVIRPPLNGDSLTEYEAGVCEDENWTDDQLLGFKPCNENLIAGCNIINGKCECNTISpecifications
NP_057525 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VCLPCDESKCEEPRNCPGSIVQGVCGCCYTCASQRNESCGGTFGIYGTCDRGLRCVIRPPLNGDSLTEYEAGVCEDENWTDDQLLGFKPCNENLIAGCNIINGKCECNTI | |
RUO | |
CRIM1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51232 | |
CRIM1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC138194/S52 | |
CRIM1 | |
Recombinant | |
wheat germ expression system |