missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CSAD Partial ORF (NP_057073, 1 a.a. - 98 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

3860.70 SEK - 5854.80 SEK

Specifications

Name CSAD (Human) Recombinant Protein (Q01)
Recombinant Yes
For Use With (Application) Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Molecular Weight 36.52kDa
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16116936
View Documents Promotion Details
Abnova™
H00051380-Q01.10UG
10 ug
3860.70 SEK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 02-03-2022
Please to purchase this item. Need a web account? Register with us today!
16126936
View Documents Promotion Details
Abnova™
H00051380-Q01.25UG
25 ug
5854.80 SEK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 02-03-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence: MADSEALPSLAGDPVAVEALLRAVFGVVVDEAIQKGTSVSQKVCEWKEPEELKQLLDLELRSQGESQKQILERCRAVIRYSVKTGHPRFFNQLFSGLD
Specifications

Specifications

CSAD (Human) Recombinant Protein (Q01)
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
36.52kDa
SDS
Product Certifications