missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CST5 Full-length ORF (NP_001891.2, 1 a.a. - 142 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001473-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193651

  • 6845.00 SEK / 25µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions. The cystatin locus on chromosome 20 contains the majority of the type 2 cystatin genes and pseudogenes. This gene is located in the cystatin locus and encodes a protein found in saliva and tears. The encoded protein may play a protective role against proteinases present in the oral cavity. [provided by RefSeq]

Sequence: MMWPMHTPLLLLTALMVAVAGSASAQSRTLAGGIHATDLNDKSVQCALDFAISEYNKVINKDEYYSRPLQVMAAYQQIVGGVNYYFNVKFGRTTCTKSQPNLDNCPFNDQPKLKEEEFCSFQINEVPWEDKISILNYKCRKV
Specifications

Specifications

NP_001891.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
42.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC71922
CST5
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
1473
CST5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MMWPMHTPLLLLTALMVAVAGSASAQSRTLAGGIHATDLNDKSVQCALDFAISEYNKVINKDEYYSRPLQVMAAYQQIVGGVNYYFNVKFGRTTCTKSQPNLDNCPFNDQPKLKEEEFCSFQINEVPWEDKISILNYKCRKV
RUO
CST5
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers