Learn More
Abnova™ Human CTCFL Partial ORF (NP_542185, 193 a.a. - 293 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_542185 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 140690 |
Molecular Weight (g/mol) | 36.85kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16181947
|
Abnova™
H00140690-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16191947
|
Abnova™
H00140690-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CCCTC-binding factor (CTCF), an 11-zinc-finger factor involved in gene regulation, utilizes different zinc fingers to bind varying DNA target sites. CTCF forms methylation-sensitive insulators that regulate X-chromosome inactivation. This gene is a paralog of CTCF and appears to be expressed primarily in the cytoplasm of spermatocytes, unlike CTCF which is expressed primarily in the nucleus of somatic cells. CTCF and the protein encoded by this gene are normally expressed in a mutually exclusive pattern that correlates with resetting of methylation marks during male germ cell differentiation. [provided by RefSeq]
Sequence: AERTKEQLFFVETMSGDERSDEIVLTVSNSNVEEQEDQPTAGQADAEKAKSTKNQRKTKGAKGTFHCDVCMFTSSRMSSFNRHMKTHTSEKPHLCHLCLKTSpecifications
NP_542185 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.85kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BORIS/CTCF-T/MGC163358/dJ579F20.2 | |
CTCFL | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
140690 | |
CTCFL (Human) Recombinant Protein (Q01) | |
AERTKEQLFFVETMSGDERSDEIVLTVSNSNVEEQEDQPTAGQADAEKAKSTKNQRKTKGAKGTFHCDVCMFTSSRMSSFNRHMKTHTSEKPHLCHLCLKT | |
RUO | |
CTCFL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |