missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human CXCR5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB190R-SP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
CXCR5 Monoclonal specifically detects CXCR5 in Human samples. It is validated for Immunohistochemistry.
Specifikationer
| CXCR5 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| BLR1, BLR1MDR-15, Burkitt lymphoma receptor 1, Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif)receptor 5), Burkitt lymphoma receptor 1, GTP-binding protein, CD185, CD185 antigen, chemokine (C-X-C motif) receptor 5, C-X-C chemokine receptor type 5, CXCR5, CXC-R5, MDR15CXCR-5, MGC117347, Monocyte-derived receptor 15 | |
| NS0 mouse myeloma cell line transfected with human CXCR5, Met1-Phe372, Accession # P32302 | |
| 25 μg | |
| 643 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Immunohistochemistry | |
| 51505R | |
| Immunohistochemistry 3-25 μg/mL | |
| P32302 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Stains human CXCR5 transfectants but not the parental cell lines in flow cytometry. | |
| Human | |
| Supernatant |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering