missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CYBB Partial ORF (AAH32720, 120 a.a. - 171 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001536-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16133801

  • 5030.41 SEK / 25µg

Estimated Shipment: 19-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cytochrome b (-245) is composed of cytochrome b alpha (CYBA) and beta (CYBB) chain. It has been proposed as a primary component of the microbicidal oxidase system of phagocytes. CYBB deficiency is one of five described biochemical defects associated with chronic granulomatous disease (CGD). In this disorder, there is decreased activity of phagocyte NADPH oxidase; neutrophils are able to phagocytize bacteria but cannot kill them in the phagocytic vacuoles. The cause of the killing defect is an inability to increase the cell's respiration and consequent failure to deliver activated oxygen into the phagocytic vacuole. [provided by RefSeq]

Sequence: LFNVEWCVNARVNNSDPYSVALSELGDRQNESYLNFARKRIKNPEGGLYLAV
Specifications

Specifications

AAH32720
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.46kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LFNVEWCVNARVNNSDPYSVALSELGDRQNESYLNFARKRIKNPEGGLYLAV
RUO
CYBB
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
1536
CYBB (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CGD/GP91-1/GP91-PHOX/GP91PHOX/NOX2/p91-PHOX
CYBB
Yes
Videos
SDS
Documents
Certificates