Learn More
Abnova™ Human CYP4F22 Full-length ORF (AAH69351.1, 1 a.a. - 531 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00126410-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This gene is part of a cluster of cytochrome P450 genes on chromosome 19 and encodes an enzyme thought to play a role in the 12(R)-lipoxygenase pathway. Mutations in this gene are the cause of ichthyosis lamellar type 3. [provided by RefSeq]
Sequence: MLPITDRLLHLLGLEKTAFRIYAVSTLLLFLLFFLFRLLLRFLRLCRSFYITCRRLRCFPQPPRRNWLLGHLGMYLPNEAGLQDEKKVLDNMHHVLLVWMGPVLPLLVLVHPDYIKPLLGASAAIAPKDDLFYGFLKPWLGDGLLLSKGDKWSRHRRLLTPAFHFDILKPYMKIFNQSADIMHAKWRHLAEGSAVSLDMFEHISLMTLDSLQKCVFSYNSNCQEKMSDYISAIIELSALSVRRQYRLHHYLDFIYYRSADGRRFRQACDMVHHFTTEVIQERRRALRQQGAEAWLKAKQGKTLDFIDVLLLARDEDGKELSDEDIRAEADTFMFEGHDTTSSGISWMLFNLAKYPEYQEKCREEIQEVMKGRELEELEWDDLTQLPFTTMCIKESLRQYPPVTLVSRQCTEDIKLPDGRIIPKGIICLVSIYGTHHNPTVWPDSKVYNPYRFDPDNPQQRSPLAYVPFSAGPRNCIGQSFAMAELRVVVALTLLRFRLSVDRTRKVRRKPELILRTENGLWLKVEPLPPRASpecifications
AAH69351.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
88.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ39501/LI3 | |
CYP4F22 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
126410 | |
CYP4F22 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLPITDRLLHLLGLEKTAFRIYAVSTLLLFLLFFLFRLLLRFLRLCRSFYITCRRLRCFPQPPRRNWLLGHLGMYLPNEAGLQDEKKVLDNMHHVLLVWMGPVLPLLVLVHPDYIKPLLGASAAIAPKDDLFYGFLKPWLGDGLLLSKGDKWSRHRRLLTPAFHFDILKPYMKIFNQSADIMHAKWRHLAEGSAVSLDMFEHISLMTLDSLQKCVFSYNSNCQEKMSDYISAIIELSALSVRRQYRLHHYLDFIYYRSADGRRFRQACDMVHHFTTEVIQERRRALRQQGAEAWLKAKQGKTLDFIDVLLLARDEDGKELSDEDIRAEADTFMFEGHDTTSSGISWMLFNLAKYPEYQEKCREEIQEVMKGRELEELEWDDLTQLPFTTMCIKESLRQYPPVTLVSRQCTEDIKLPDGRIIPKGIICLVSIYGTHHNPTVWPDSKVYNPYRFDPDNPQQRSPLAYVPFSAGPRNCIGQSFAMAELRVVVALTLLRFRLSVDRTRKVRRKPELILRTENGLWLKVEPLPPRA | |
RUO | |
CYP4F22 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |