Learn More
Abnova™ Human CYTH3 Full-length ORF (AAH08191.1, 1 a.a. - 179 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH08191.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9265 |
Molecular Weight (g/mol) | 45.43kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16183472
|
Abnova™
H00009265-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16173472
|
Abnova™
H00009265-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the PSCD (pleckstrin homology, Sec7 and coiled-coil domains) family. PSCD family members have identical structural organization that consists of an N-terminal coiled-coil motif, a central Sec7 domain, and a C-terminal pleckstrin homology (PH) domain. The coiled-coil motif is involved in homodimerization, the Sec7 domain contains guanine-nucleotide exchange protein (GEP) activity, and the PH domain interacts with phospholipids and is responsible for association of PSCDs with membranes. Members of this family appear to mediate the regulation of protein sorting and membrane trafficking. This encoded protein is involved in the control of Golgi structure and function, and it may have a physiological role in regulating ADP-ribosylation factor protein 6 (ARF) functions, in addition to acting on ARF1. [provided by RefSeq]
Sequence: MNRGINEGGDLPEELLRNLYESIKNEPFKIPEDDGNDLTHTFFNPDREGWLLKLGGRVKTWKRRWFILTDNCLYYFEYTTDKEPRGIIPLENLSIREVEDPRKPNCFELYNPSHKGQVIKACKTEADGRVVEGNHVVYRISAPSPEEKEEWMKSIKASISRDPFYDMLATRKRRIANKKSpecifications
AAH08191.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45.43kDa | |
Glutathione Sepharose 4 Fast Flow | |
MNRGINEGGDLPEELLRNLYESIKNEPFKIPEDDGNDLTHTFFNPDREGWLLKLGGRVKTWKRRWFILTDNCLYYFEYTTDKEPRGIIPLENLSIREVEDPRKPNCFELYNPSHKGQVIKACKTEADGRVVEGNHVVYRISAPSPEEKEEWMKSIKASISRDPFYDMLATRKRRIANKK | |
RUO | |
CYTH3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9265 | |
CYTH3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARNO3/GRP1/PSCD3 | |
CYTH3 | |
Recombinant | |
wheat germ expression system |