missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DAO Partial ORF (NP_001908, 119 a.a. - 218 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001610-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16177014

  • 3343.96 SEK / 10µg

Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes the peroxisomal enzyme D-amino acid oxidase. The enzyme is a flavoprotein which uses flavin adenine dinucleotide (FAD) as its prosthetic group. Its substrates include a wide variety of D-amino acids, but it is inactive on the naturally occurring L-amino acids. Its biological function is not known; it may act as a detoxifying agent which removes D-amino acids that accumulate during aging. In mice, it degrades D-serine, a co-agonist of the NMDA receptor. This gene may play a role in the pathophysiology of schizophrenia. [provided by RefSeq]

Sequence: PRELDMFPDYGYGWFHTSLILEGKNYLQWLTERLTERGVKFFQRKVESFEEVAREGADVIVNCTGVWAGALQRDPLLQPGRGQIMKVDAPWMKHFILTHD
Specifications

Specifications

NP_001908
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PRELDMFPDYGYGWFHTSLILEGKNYLQWLTERLTERGVKFFQRKVESFEEVAREGADVIVNCTGVWAGALQRDPLLQPGRGQIMKVDAPWMKHFILTHD
RUO
DAO
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
1610
DAO (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DAAO/DAMOX/MGC35381/OXDA
DAO
Yes
Videos
SDS
Documents
Certificates