Learn More
Abnova™ Human DCAMKL1 Partial ORF (NP_004725, 640 a.a. - 729 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009201-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
DCAMKL1 is a microtubule-associated protein that phosphorylates itself and myelin basic protein (MBP; MIM 159430). DCAMKL1 has microtubule polymerizing activity that is independent of its protein kinase activity (Lin et al., 2000 [PubMed 11124993]).[supplied by OMIM]
Sequence: QVLEHPWVNDDGLPENEHQLSVAGKIKKHFNTGPKPNSTAAGVSVIALDHGFTIKRSGSLDYYQQPGMYWIRPPLLIRRGRFSDEDATRMSpecifications
NP_004725 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QVLEHPWVNDDGLPENEHQLSVAGKIKKHFNTGPKPNSTAAGVSVIALDHGFTIKRSGSLDYYQQPGMYWIRPPLLIRRGRFSDEDATRM | |
RUO | |
DCLK1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9201 | |
DCAMKL1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DCAMKL1/DCDC3A/DCLK/KIAA0369 | |
DCLK1 | |
Recombinant | |
wheat germ expression system |