Learn More
Abnova™ Human DCTN2 Full-length ORF (AAH00718.1, 1 a.a. - 401 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH00718.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10540 |
Molecular Weight (g/mol) | 70.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16185272
|
Abnova™
H00010540-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16175272
|
Abnova™
H00010540-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a 50-kD subunit of dynactin, a macromolecular complex consisting of 10-11 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit is present in 4-5 copies per dynactin molecule. It contains three short alpha-helical coiled-coil domains that may mediate association with self or other dynactin subunits. It may interact directly with the largest subunit (p150) of dynactin and may affix p150 in place. [provided by RefSeq]
Sequence: MADPKYADLPGIARNEPDVYETSDLPEDDQAEFDAEELTSTSVEHIIVNPNAAYDKFKDKRVGTKGLDFSDRIGKTKRTGYESGEYEMLGEGLGVKETPQQKYQRLLHEVQELTTEVEKIKTTVKESATEEKLTPVLLAKQLAALKQQLVASHLEKLLGPDAAINLTDPDGALAKRLLLQLEATKNSKGGSGGKTTGTPPDSSLVTYELHSRPEQDKFSQAAKVAELEKRLTELETAVRCDQDAQNPLSAGLQGACLMETVELLQAKVSALDLAVLDQVEARLQSVLGKVNEIAKHKASVEDADTQSKVHQLYETIQRWSPIASTLPELVQRLVTIKQLHEQAMQFGQLLTHLDTTQQMIANSLKDNTTLLTQVQTTMRENLATVEGNFASIDERMKKLGKSpecifications
AAH00718.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
70.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MADPKYADLPGIARNEPDVYETSDLPEDDQAEFDAEELTSTSVEHIIVNPNAAYDKFKDKRVGTKGLDFSDRIGKTKRTGYESGEYEMLGEGLGVKETPQQKYQRLLHEVQELTTEVEKIKTTVKESATEEKLTPVLLAKQLAALKQQLVASHLEKLLGPDAAINLTDPDGALAKRLLLQLEATKNSKGGSGGKTTGTPPDSSLVTYELHSRPEQDKFSQAAKVAELEKRLTELETAVRCDQDAQNPLSAGLQGACLMETVELLQAKVSALDLAVLDQVEARLQSVLGKVNEIAKHKASVEDADTQSKVHQLYETIQRWSPIASTLPELVQRLVTIKQLHEQAMQFGQLLTHLDTTQQMIANSLKDNTTLLTQVQTTMRENLATVEGNFASIDERMKKLGK | |
RUO | |
DCTN2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10540 | |
DCTN2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DCTN50/DYNAMITIN/RBP50 | |
DCTN2 | |
Recombinant | |
wheat germ expression system |